ContactUsLinkComponent
Cynomolgus DNAM-1, His Tag (DN1-C52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Phe 252 (Accession A0A2K5 W1Z6-1). 1 * 100 µG
Catalog # ACRBDN1-C52H9100UG
Supplier:  ACROBIOSYSTEMS
Cynomolgus DNAM-1, His Tag (DN1-C52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Phe 252 (Accession A0A2K5 W1Z6-1). 1 * 100 µG
Catalog # ACRBDN1-C52H9100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together