ContactUsLinkComponent
Biotinylated Cynomolgus CD64, His,Avitag (BLI verified) (FCA-C82E8) is expressed from human 293 cells (HEK293). It contains AA Val 1 1 - Pro 288 (Accession NP001270969.1). 1 * 25 µG
Catalog # ACRBFCA-C82E8-25UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus CD64, His,Avitag (BLI verified) (FCA-C82E8) is expressed from human 293 cells (HEK293). It contains AA Val 1 1 - Pro 288 (Accession NP001270969.1). 1 * 25 µG
Catalog # ACRBFCA-C82E8-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together