ContactUsLinkComponent
Biotinylated Human IL-12 R beta 1, Fc,Avitag (ILB-H82F7) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 540 (Accession P42701-1). 1 * 200 µG
  ACRBILB-H82F7200UG
Biotinylated Human IL-12 R beta 1, Fc,Avitag (ILB-H82F7) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 540 (Accession P42701-1). 1 * 200 µG
  ACRBILB-H82F7200UG
Frequently Bought Together