ContactUsLinkComponent
Biotinylated Human CD32b, Avitag,His Tag (SPR  BLI verified) (CDB-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Accession P31994-1). 1 * 25 µG
Catalog # ACRBCDB-H82E0-25UG
Supplier:  ACRO Biosystems
Biotinylated Human CD32b, Avitag,His Tag (SPR  BLI verified) (CDB-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Accession P31994-1). 1 * 25 µG
Catalog # ACRBCDB-H82E0-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together