ContactUsLinkComponent
Human TIM-3, Mouse IgG2a Fc Tag, low endotoxin (TM3-H525b) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 2 00 (Accession Q8TDQ0-1). 1 * 100 µG
  ACRBTM3-H525B100UG
Human TIM-3, Mouse IgG2a Fc Tag, low endotoxin (TM3-H525b) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 2 00 (Accession Q8TDQ0-1). 1 * 100 µG
  ACRBTM3-H525B100UG
Frequently Bought Together