ContactUsLinkComponent
Human TSLP (R127A, R130A), His Tag (TSP-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accessio n Q969D9-1 (R127A, R130A)). 1 * 100 µG
Catalog # ACRBTSP-H52HA100UG
Supplier:  ACRO Biosystems
Human TSLP (R127A, R130A), His Tag (TSP-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accessio n Q969D9-1 (R127A, R130A)). 1 * 100 µG
Catalog # ACRBTSP-H52HA100UG
Supplier:  ACRO Biosystems
Frequently Bought Together