ContactUsLinkComponent
Cynomolgus Complement C5, His Tag (CO5-C52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accessio n XP005580972.1). 1 * 100 µG
Catalog # ACRBCO5-C52HX100UG
Supplier:  ACRO Biosystems
Cynomolgus Complement C5, His Tag (CO5-C52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accessio n XP005580972.1). 1 * 100 µG
Catalog # ACRBCO5-C52HX100UG
Supplier:  ACRO Biosystems
Frequently Bought Together