Human Complement C5 (R885H), His Tag (CO5-H52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Acces sion P01031-1 (R885H)). 1 * 1 mg
Catalog # ACRBCO5-H52HX-1MG
Supplier: ACRO Biosystems
Frequently Bought Together