ContactUsLinkComponent
Human LIF R, Fc Tag (LIR-H4252) is expressed from human 293 cells (HEK293). It contains AA Gln 45 - Ser 833 (Accession P42702-1). 1 * 50 µG
Catalog # ACRBLIR-H4252-50UG
Supplier:  ACRO Biosystems
Human LIF R, Fc Tag (LIR-H4252) is expressed from human 293 cells (HEK293). It contains AA Gln 45 - Ser 833 (Accession P42702-1). 1 * 50 µG
Catalog # ACRBLIR-H4252-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together