ContactUsLinkComponent
CP-Wax 52 CB, 60 m, 0.25 mm, 0.25 μm GC column, 7 inch cage 1 * 1 SET
Catalog # HEWLCP8723I
Supplier:  Agilent
undefined
CP-Wax 52 CB, 60 m, 0.25 mm, 0.25 μm GC column, 7 inch cage 1 * 1 SET
Catalog # HEWLCP8723I
Supplier:  Agilent
Frequently Bought Together