ContactUsLinkComponent
Semi-automatic, electro mechanically controlled benchtop laboratory autoclave with EU-plug, Chamberdimension (ØxL) 250 x 420 mm Volume: 23 liters 1 * 1 items
Catalog # TUTT2540ML
Supplier:  TUTTNAUER
Semi-automatic, electro mechanically controlled benchtop laboratory autoclave with EU-plug, Chamberdimension (ØxL) 250 x 420 mm Volume: 23 liters 1 * 1 items
Catalog # TUTT2540ML
Supplier:  TUTTNAUER
Frequently Bought Together