Bag waste disposal, 72l , biohazard ,with indicator field , 700 x 1100 x 0.050 mm 1 * 75 items
Catalog # RATI7002305
Supplier: RATIOLAB
Bag waste disposal, 72l , biohazard ,with indicator field , 700 x 1100 x 0.050 mm 1 * 75 items
Catalog # RATI7002305
Supplier: RATIOLAB
Frequently Bought Together
VWR® Disposable Spatulas, PP
Supplier: VWR Collection
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Autoclave Sterilisation Tape
Supplier: KARTELL
epT.I.P.S.® Reloads, Pipette Tips
Supplier: EPPENDORF
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
VWR®, Pipette Tips, Universal
Supplier: VWR Collection
Pouring rings for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Pipette Tips, Standard
Supplier: VWR Collection
VWR®, Biohazard bags with or without sterilisation indicator strip
Supplier: VWR Collection
epT.I.P.S.® Bulk - Pipette Tips
Supplier: EPPENDORF
VWR®, Forceps
Supplier: VWR Collection
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
Supplier: EPPENDORF
VWR® Reagent Reservoirs
Supplier: VWR Collection