Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cryo-Boxes, 1/4-Format, PC, grid 5 x 5, blue, 76 x 76 x 52 mm 1 * 8 items
Catalog # RATI5150031
Supplier:
RATIOLAB
Expand image
Cryo-Boxes, 1/4-Format, PC, grid 5 x 5, blue, 76 x 76 x 52 mm 1 * 8 items
Catalog # RATI5150031
Supplier:
RATIOLAB
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Cell culture flasks, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection