1 component Benzene [CAS:71-43-2] 250mg/l in Methanol . Duree de conservation : 12 mois. Store in a refrigerator at temperatures between 2C to 8C 1 * 10 mL
Catalog # CMPLRD03720410.5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together