Reflector module C-DIC/TIC ACR P&C for reflected light optimized for circular DIC in reflected light, for use with C-DIC/ TIC sliders 6x20, not suitable for polarization 1 * 1 ST
Catalog # ZEIS4249419000000
Supplier: ZEISS
Frequently Bought Together