ContactUsLinkComponent
10 µg-The Thermo Scientific pMCS Tluc16-DD Vector is a multiple cloning site (MCS) vector designed to accept a promoter sequence for study of gene regulation using the intracellular TurboLuc16 (Tluc16) luciferase reporter. 1 * 10 µG
Catalog # PIER88231
10 µg-The Thermo Scientific pMCS Tluc16-DD Vector is a multiple cloning site (MCS) vector designed to accept a promoter sequence for study of gene regulation using the intracellular TurboLuc16 (Tluc16) luciferase reporter. 1 * 10 µG
Catalog # PIER88231
Frequently Bought Together