Orders LinkContactUsLinkComponent
Sample vessel
Sample vessel
25 ml, Sample vessels
Catalog # HACHLZQ035
Supplier:   Hach
Sample vessel
Catalog # HACHLZQ035
Supplier:  Hach
Sample vessel
Sample vessel
25 ml, Sample vessels
Price
Inquire For Price
Log in for availability
1 * 200 items
Product Details & Documents
Sample vessel made from PS. For aqueous solutions, acids and bases.
For use with autosampler LPV122/LZQ031.
Specifications
  • Capacity:
    25 ml
  • Description:
    Sample vessels