ContactUsLinkComponent
Handheld instrument for gaseous oxygen with datalogger GMH3695 Set-Option: Device only, without sensor Measuring range oxygen: 0.0 ... 100.0 % O. Vol Measuring range temperature: -5 ... +50 #‹C Measuring range air pressure: 300 ... 5000 hPa abs, pres 1 * 1 items
Catalog # GRSG605921
Supplier:  Greisinger
Handheld instrument for gaseous oxygen with datalogger GMH3695 Set-Option: Device only, without sensor Measuring range oxygen: 0.0 ... 100.0 % O. Vol Measuring range temperature: -5 ... +50 #‹C Measuring range air pressure: 300 ... 5000 hPa abs, pres 1 * 1 items
Catalog # GRSG605921
Supplier:  Greisinger
Frequently Bought Together