Orders LinkContactUsLinkComponent
Horizontal fold flat disposable masks, Springfit™
Horizontal fold flat disposable masks, Springfit™
Springfit™ Typhoon™ Disposable mask, valved, FFP3
Catalog # JSPABGA802-206-000
Supplier:   JSP
Horizontal fold flat disposable masks, Springfit™
Catalog # JSPABGA802-206-000
Supplier:  JSP
Horizontal fold flat disposable masks, Springfit™
Horizontal fold flat disposable masks, Springfit™
Springfit™ Typhoon™ Disposable mask, valved, FFP3
Price
57,40 €
Log in for availability
1 * 10 items
Product Details & Documents
The SpringFit™ fold flat disposable mask achieves an unrivalled secure fit with the use of a unique endoskeleton structure. SpringFit™ introduces an abundance of features that maximise fit, comfort, compatibility, and performance.
  • Fold flat disposable mask
  • Compact and easily pocketed
  • Unique endoskeleton structure
  • Polypropylene inner skeleton
  • Available in FFP3 and FFP2, valved and unvalved
The SpringFit™ fold flat disposable mask is available in FFP3 and FFP2, valved and unvalved, with a nuisance odour carbon layer available on the FFP3 model. The Unique Typhoon™ valve has been designed to reduce breathing resistance, heat, and moisture build-up. The valve is unaffected by any moisture, and therefore can be used with the same performance under very low temperatures.
The fold flat design makes SpringFit™ more compact, and easily pocketed. SpringFit™ is also individually packed to ensure a high level of hygiene.
Certifications: EN149:2001+A1:2009.
Specifications
  • Description:
    Springfit™ Typhoon™ Disposable mask, valved
  • Seq:
    05
  • Class:
    FFP3
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.