Safety spectacles, Eiger™
Eiger™ safety spectacles, Black/green, Smoke
Catalog # JSPA1EIGGREEN23SKN
Supplier: JSP
Jump to:
Product Details & Documents
Sporty design with optimal curved lens and large field of vision.
- Moulded browguard
- Excellence coating
- Optimal lateral coverage
Active style and sporty design. Unique Flexofit technology engineered into temple cushioning to enhance wearer comfort, and to ensure a non-slip, low pressure fit.
Excellence coating on the optimal curved lens, large field of vision, moulded browguard. Additional gasket and strap accessory pack available to deliver a perfect dust barrier.
: Meets EN 166:2001.
Specifications
- Colour:Black/green
- Description:Eiger™ safety spectacles
- Seq:02
- Lenses:Smoke