Horizontal fold flat disposable masks, Springfit™
Product Details & Documents
The SpringFit™ fold flat disposable mask achieves an unrivalled secure fit with the use of a unique endoskeleton structure. SpringFit™ introduces an abundance of features that maximise fit, comfort, compatibility, and performance.
- Fold flat disposable mask
- Compact and easily pocketed
- Unique endoskeleton structure
- Polypropylene inner skeleton
- Available in FFP3 and FFP2, valved and unvalved
The SpringFit™ fold flat disposable mask is available in FFP3 and FFP2, valved and unvalved, with a nuisance odour carbon layer available on the FFP3 model. The Unique Typhoon™ valve has been designed to reduce breathing resistance, heat, and moisture build-up. The valve is unaffected by any moisture, and therefore can be used with the same performance under very low temperatures.
The fold flat design makes SpringFit™ more compact, and easily pocketed. SpringFit™ is also individually packed to ensure a high level of hygiene.
Certifications: EN149:2001+A1:2009.
Specifications
- Description:Springfit™ Disposable mask, unvalved
- Seq:02
- Class:FFP3