ContactUsLinkComponent
Draft ring for Sartorius Entris® analytical balances. For Sartorius lab balances of the product family Entris® with a readability of 0.1 mg. Scope of delivery: - Operation instruction 1 * 1 items
Catalog # SARBYSP02
Supplier:  Sartorius Balances
Draft ring for Sartorius Entris® analytical balances. For Sartorius lab balances of the product family Entris® with a readability of 0.1 mg. Scope of delivery: - Operation instruction 1 * 1 items
Catalog # SARBYSP02
Supplier:  Sartorius Balances
Frequently Bought Together