Orders LinkContactUsLinkComponent
Objectives for microscopes, Panthera TEC POL series
Objectives for microscopes, Panthera TEC POL series
CCIS® Plan achromatic strain-free objective UC PL P 4×/0,10 (WD=30,5 mm)
Catalog # MOTI1101001707191
Supplier:   MOTIC
Objectives for microscopes, Panthera TEC POL series
Catalog # MOTI1101001707191
Supplier:  MOTIC
Objectives for microscopes, Panthera TEC POL series
Objectives for microscopes, Panthera TEC POL series
CCIS® Plan achromatic strain-free objective UC PL P 4×/0,10 (WD=30,5 mm)
Price
105,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    CCIS® Plan achromatic strain-free objective UC PL P 4×/0,10 (WD=30,5 mm)
  • Seq:
    01