Orders LinkContactUsLinkComponent
Objectives for microscopes, Panthera TEC POL series
CCIS® Plan achromatic strain-free objective UC PL P 40×/0,65/S (WD=0,6 mm)
  MOTI1101001707221
 :   MOTIC
Objectives for microscopes, Panthera TEC POL series
  MOTI1101001707221
 :  MOTIC
Objectives for microscopes, Panthera TEC POL series
Objectives for microscopes, Panthera TEC POL series
CCIS® Plan achromatic strain-free objective UC PL P 40×/0,65/S (WD=0,6 mm)
357,00 €
1 * 1 items
  • Description:
    CCIS® Plan achromatic strain-free objective UC PL P 40×/0,65/S (WD=0,6 mm)
  • Seq:
    04