Volumetric flask, clear glass, 50 ml, NS 14/23, class A, conformity certified, ringmark, DIFFICO brown graduation, with ground stoppers bed, without stopper 1 * 1 items
Catalog # WITG3.662.006
Supplier: witeg Labortechnik
Frequently Bought Together