Photo hood phone platform MiniOne. Adapter connects to the vents of the photo hood and creates a platform to rest a mobile device on for documenting gels and taking time lapse videos. 1 * 5 items
EMBIM2017
: EMBITEC
Frequently Bought Together