ContactUsLinkComponent
MBT Galaxy HCCA Matrix GPR Purified matrix substance for MALDI Biotyper™ Galaxy automated target preparation system alpha-cyano-4-hy droxycinnamic acid C10H7NO3 M 189.17 - CAS 28166-41-8 Content: 10 tubes à 7.5 mg HCCA 1 * 10 Vial
Catalog # MSPP1823405
MBT Galaxy HCCA Matrix GPR Purified matrix substance for MALDI Biotyper™ Galaxy automated target preparation system alpha-cyano-4-hy droxycinnamic acid C10H7NO3 M 189.17 - CAS 28166-41-8 Content: 10 tubes à 7.5 mg HCCA 1 * 10 Vial
Catalog # MSPP1823405