ContactUsLinkComponent
PAS -MAYERS HEMATOXILYN IS USED TO DEMONSTRATE SIMPLE POLYSACCHARIDES, NEUTRAL MUCO-POLYSACCHARIDES, MUCOPROTEINS, AND GLYCOLIPIDS. 1 * 500 mL
Catalog # GENOBAQ090
Supplier:  G-Biosciences
PAS -MAYERS HEMATOXILYN IS USED TO DEMONSTRATE SIMPLE POLYSACCHARIDES, NEUTRAL MUCO-POLYSACCHARIDES, MUCOPROTEINS, AND GLYCOLIPIDS. 1 * 500 mL
Catalog # GENOBAQ090
Supplier:  G-Biosciences
Frequently Bought Together