ContactUsLinkComponent
Glass Microfibre Filters, grade MGA sheets: 203 x 254 mm Packing unit: 100 pieces Properties: Basis weight: 52 g/m² Air resistance: 36 mbar Thickness: 0.25 mm Particle retention: 1.6 µm 1 * 100 items
Catalog # SARTFT21101203254
Supplier:  Sartorius
Glass Microfibre Filters, grade MGA sheets: 203 x 254 mm Packing unit: 100 pieces Properties: Basis weight: 52 g/m² Air resistance: 36 mbar Thickness: 0.25 mm Particle retention: 1.6 µm 1 * 100 items
Catalog # SARTFT21101203254
Supplier:  Sartorius
Frequently Bought Together