ContactUsLinkComponent
CX23LEDRFS1-1-3 microscope set Upright microscope set for transmitted light observation. CX23 microscope frame with integrated LED illumination, quadruple nosepiece, fixed mechanical stage (120 x 132 mm, travelling range 76 x 30 mm) with right hand l 1 * 1 items
Catalog # OLYMN5731000
Supplier:  OLYMPUS OPTICAL
CX23LEDRFS1-1-3 microscope set Upright microscope set for transmitted light observation. CX23 microscope frame with integrated LED illumination, quadruple nosepiece, fixed mechanical stage (120 x 132 mm, travelling range 76 x 30 mm) with right hand l 1 * 1 items
Catalog # OLYMN5731000
Supplier:  OLYMPUS OPTICAL
Frequently Bought Together