SC2-P3496-A PolyGard®-2 Sensor Cartridge, 5VDC Benzindämpfe:0-100 %UEG (Pellistorsensor) zum Anschluss an SB2/MSB2/MSC2, digitale Kalibration,Schutzart:IP65 1 * 1 ST
Catalog # SNSTSC2-P3496-A
Supplier: SENSORTECHNIK TRENKA
Frequently Bought Together