ContactUsLinkComponent
Aldrich(R) rectangular TLC developing tanks, complete, L x H x W 27.0 cm x 26.5 cm x 7.0 cm 1 * 1 items
Catalog # SIALZ126195-1EA
Supplier:  Merck
Aldrich(R) rectangular TLC developing tanks, complete, L x H x W 27.0 cm x 26.5 cm x 7.0 cm 1 * 1 items
Catalog # SIALZ126195-1EA
Supplier:  Merck
Frequently Bought Together