ContactUsLinkComponent
Second Source of Oregon Residual Solvent Mix, 35 Components, 1000 µg/mL in Triacetin 1 * 1 mL
Catalog # EHERGA09000238TNSS
Supplier:  EHRENSTORFER
Second Source of Oregon Residual Solvent Mix, 35 Components, 1000 µg/mL in Triacetin 1 * 1 mL
Catalog # EHERGA09000238TNSS
Supplier:  EHRENSTORFER
Frequently Bought Together