Tri-Clamp GL-Adaptor, Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to a GL-threaded neck. Universal chemical r esistance, the flowing product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLD901-06
Supplier: Bohlender
Tri-Clamp GL-Adaptor, Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to a GL-threaded neck. Universal chemical r esistance, the flowing product is only exposed to PTFE. 1 * 1 items
Catalog # BOHLD901-06
Supplier: Bohlender
Frequently Bought Together
VWR®, Measuring Beakers with Handle, PP
Supplier: VWR Collection
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
epT.I.P.S.® Reloads, Pipette Tips
Supplier: EPPENDORF
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF
Autoclavable Bags, Dispenser Box
Supplier: RATIOLAB
epT.I.P.S.® Bulk - Pipette Tips
Supplier: EPPENDORF
VWR®, Forceps
Supplier: VWR Collection
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
VWR®, Disposable Weighing Boats
Supplier: VWR Collection
Pouring rings for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Supplier: EPPENDORF
VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
VWR® Reagent Reservoirs
Supplier: VWR Collection
Threaded couplings
Supplier: Bohlender
Joint clamps for conical ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
Tubings, PFA, BOLA
Supplier: Bohlender
BOLA FLASCHEN-MEHRFACHVERTEILER, PTFE
Supplier: Bohlender