ContactUsLinkComponent
ACQUITY UPLC Protein BEH C4 Column, 300Å, 1.7 µm, 1 mm X 150 mm 1 * 1 items
Catalog # WATE186005591
Supplier:  WATERS
ACQUITY UPLC Protein BEH C4 Column, 300Å, 1.7 µm, 1 mm X 150 mm 1 * 1 items
Catalog # WATE186005591
Supplier:  WATERS
Frequently Bought Together