NTRK3 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human TRKC in cell culture sup ernates, serum and plasma (heparin, EDTA). 1 * 1 KIT
NTRK3 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human TRKC in cell culture sup ernates, serum and plasma (heparin, EDTA). 1 * 1 KIT