ContactUsLinkComponent
IL1F10 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human IL-38 in cell culture s upernates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5458
Supplier:  Abnova
IL1F10 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human IL-38 in cell culture s upernates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5458
Supplier:  Abnova
Frequently Bought Together