ContactUsLinkComponent
Glass Microfibre Filters, grade MG 550-HA Filter discs, diameter: 47 mm Packing unit: 100 pieces Basis weight: 65 g/m² Thickness: 0.30 mm Particle retention: 1.5 µm Penetration 0,3 µm: 0,02 % Temperature resistant up to 550 °C 1 * 100 items
Catalog # SARTFT-3-01147-047
Supplier:  Sartorius
Glass Microfibre Filters, grade MG 550-HA Filter discs, diameter: 47 mm Packing unit: 100 pieces Basis weight: 65 g/m² Thickness: 0.30 mm Particle retention: 1.5 µm Penetration 0,3 µm: 0,02 % Temperature resistant up to 550 °C 1 * 100 items
Catalog # SARTFT-3-01147-047
Supplier:  Sartorius
Frequently Bought Together