ContactUsLinkComponent
Mouse 4-1BB Ligand, Fc Tag (41L-M5257) is expressed from human 293 cells (HEK293). It contains AA Arg 104 - Glu 309 (Accession NP033 430). 1 * 50 µG
Catalog # ACRB41L-M5257-50UG
Supplier:  ACRO Biosystems
Mouse 4-1BB Ligand, Fc Tag (41L-M5257) is expressed from human 293 cells (HEK293). It contains AA Arg 104 - Glu 309 (Accession NP033 430). 1 * 50 µG
Catalog # ACRB41L-M5257-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together