Orders LinkContactUsLinkComponent
Assistent® Pipette brushes
Assistent® Pipette brushes
110 mm, 8 mm, 500 mm
Catalog # HECH91120000008
Assistent® Pipette brushes
Catalog # HECH91120000008
Assistent® Pipette brushes
Assistent® Pipette brushes
110 mm, 8 mm, 500 mm
Price
398,00 €
Log in for availability
1 * 50 items
Product Details & Documents
Straight brushes in different sizes.
Documents
Specifications
  • Brush head Ø:
    8 mm
  • Seq:
    08
  • Total length:
    500 mm
  • Brush head length:
    110 mm