Orders LinkContactUsLinkComponent
BD Eclipse™ Needles
BD Eclipse™ Needles
Needle, sterile, 25 G, 16 mm
Catalog # BDAM305760
Supplier:   BD Medical
BD Eclipse™ Needles
Catalog # BDAM305760
Supplier:  BD Medical
BD Eclipse™ Needles
BD Eclipse™ Needles
Needle, sterile, 25 G, 16 mm
Price
43,20 €
Log in for availability
1 * 100 items
Product Details & Documents
The BD Eclipse™ SmartSlip™ is a single use sterile needle with a safety mechanism that covers the needle point after use. In the activated position, the needle cover guards against accidental needlestick during normal handling and disposal of the used needle/syringe.
  • The SmartSlip™ Technology is specially designed for safe assembling of BD Eclipse™ needles to Luer slip syringes
  • Bevel oriented to the safety cover to allow low angle injections
  • By assembling BD Eclipse™ needles to a luer slip syringe, an audible click indicates a safely assembled needle
  • Safety can be activated immediately after injection
  • Wide, textured finger pad for thumb or forefinger
  • Single-handed with one finger activation of the safety shield, audible click indicates that the cover has safely engaged the needle
  • Natural Rubber Latex is not part of the material formulation
  • Sterile, single use
The SmartSlip™ Technology consists of an adaptor to enable connection of the BD Eclipse safety needle to a Luer slip.
Specifications
  • Gauge:
    25 G
  • Description:
    Needle, sterile
  • Length:
    16 mm
  • Seq:
    05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.