ContactUsLinkComponent
Dispenser Ceramus 10-60MlPiston made of high-purity aluminium oxide ceramic, Threaded suction hose fitting, steam sterilised at 121 ° C without dismantling 1 * 1 items
Catalog # HIRS9362001
Supplier:  HIRSCHMANN
Dispenser Ceramus 10-60MlPiston made of high-purity aluminium oxide ceramic, Threaded suction hose fitting, steam sterilised at 121 ° C without dismantling 1 * 1 items
Catalog # HIRS9362001
Supplier:  HIRSCHMANN
Frequently Bought Together