Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Dispenser ceramus 0.2-1.0ml conformity certifiedPiston made of high-purity aluminium oxide ceramic, Threaded suction hose fitting, steam sterilised at 121° C without dismantling 1 * 1 items
HIRS9312001
:
HIRSCHMANN
Dispenser ceramus 0.2-1.0ml conformity certifiedPiston made of high-purity aluminium oxide ceramic, Threaded suction hose fitting, steam sterilised at 121° C without dismantling 1 * 1 items
HIRS9312001
:
HIRSCHMANN
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals