BATH ARMOR 12 LITRE PACK 1 * 12 L
Catalog # NICKBA-12
Supplier: NICKEL ELECTRO
Frequently Bought Together
Distance sieve ZM 300 titanium trapezoid holes 2.00 mm 1 * 1 items
Supplier: Retsch GmbH
Magnetic Stirrers with Remote Control, Multi-Position
Supplier: THERMO H + P LABORTECHNIK
Magnetic Stirrers, Hei-PLATE Mix 'n' Heat Expert
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for Optima™ heated circulating baths
Supplier: GRANT INSTRUMENTS
Racks for orbital shakers SM 30 A, SM 30 A control, SM 30 B and SM 30 B control
Supplier: Edmund Bühler GmbH
Pierceable septums, pack of 10 pcs 1 * 10 items
Supplier: VELP SCIENTIFIC
VWR®, Multi Purpose Clamps, 3-Prong
Supplier: VWR Collection
Accessories for VWR® Series Homogenisers
Supplier: VWR Collection
DISPERSING TOOL S 25 EC-T-C-25G-ST 1 * 1 items
Supplier: IKA
Accessories for reciprocating shakers, HS 260
Supplier: IKA
Accessories for PURA™ water baths
Supplier: JULABO GmbH
Accessories for Qsonica sonicators
Supplier: QSONICA LLC
Accessories for laboratory mixers
Supplier: WARING BLENDOR