ContactUsLinkComponent
HPLC guard column EC 4/3 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 4 mm, ID: 3 mm pack of 3 - require Column Protection System gu ard column holder (REF 718966) - for analytical columns with 3, 4 or 4.6 mm ID 1 * 1 items
Catalog # MANA763438.30
Supplier:  MACHEREY-NAGEL
HPLC guard column EC 4/3 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 4 mm, ID: 3 mm pack of 3 - require Column Protection System gu ard column holder (REF 718966) - for analytical columns with 3, 4 or 4.6 mm ID 1 * 1 items
Catalog # MANA763438.30
Supplier:  MACHEREY-NAGEL
Frequently Bought Together