HPLC guard column EC 4/2 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 4 mm, ID: 2 mm pack of 3 - require Column Protection System gu ard column holder (REF 718966) - for analytical columns with 2 mm ID 1 * 1 items
Catalog # MANA763438.20
Supplier: MACHEREY-NAGEL
Frequently Bought Together