ContactUsLinkComponent
J&W CP-Wax 52 CB, 60 m, 0.32 mm, 0.25 µm, GC column, 7 inch cage 1 * 1 items
Catalog # HEWLCP8853I
Supplier:  Agilent
J&W CP-Wax 52 CB, 60 m, 0.32 mm, 0.25 µm, GC column, 7 inch cage 1 * 1 items
Catalog # HEWLCP8853I
Supplier:  Agilent
Frequently Bought Together