Orders LinkContactUsLinkComponent
Accessories for circulators
Accessories for circulators
3-way header, M38×1,5, The 3-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.
Catalog # HUBE343234
Supplier:   HUBER
Accessories for circulators
Catalog # HUBE343234
Supplier:  HUBER
Accessories for circulators
3-way header, M38×1,5, The 3-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.
Price
600,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    3-way header, M38×1,5
  • Seq:
    11
  • For:
    The 3-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.