D10 EJECTOR SPACER FOR PIPETMAN M 8X 1 * 1 items
Catalog # GILSF807117
Supplier: GILSON
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Accessories for PIPETBOY acu / pro
Supplier: Integra
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Cover gasket 1 * 1 items
Supplier: VWR Collection
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for accu-jet® pro
Supplier: Brand
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG