ProSpect C. difficile Toxin A/B Microplate Assay is a qualitative enzyme immunoassay (EIA) for the detection of C. difficile Toxin A and B in human faecal specimens from patients suspected of having Clostridium difficile disease. The test is intende 1 * 1 KIT
OXOIR244596
Frequently Bought Together